Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + c-MER Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + c-MER Kinase Enzyme System, 1 each, Promega
Catalog # PAV9561
CAS Number:  
undefined
ADP-Glo Kinase Assay + c-MER Kinase Enzyme System, 1 each, Promega
Catalog # PAV9561
Supplier Number:  V9561
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + c-MER Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9561
  • Supplier no.:
    V9561

Specifications

About this item

Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family.

  • Easily Screen and Profile c-MER Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family. The c-MER gene encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type domains and one tyrosine kinase domain. Functional knockout of c-MER in mice shows the mice to have macrophages deficient in the clearance of apoptotic thymocytes thereby demonstrating the critical role for c-MER in the engulfment and efficient clearance of apoptotic cells. Defects in c-MER have been associated with retinitis pigmentosa, a phenotype that can be reversed by replacement of the c-MER gene product. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: c-MER, 10ug (Human, recombinant; amino acids 578-872). MW: ~58kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. c-MER NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/10461/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.