Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4057
CAS Number:  
undefined
ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each, Promega
Catalog # PAV4057
Supplier Number:  V4057
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4057
  • Supplier no.:
    V4057

Specifications

About this item

Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases.

  • Easily Screen and Profile CLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases. The phosphorylated serine/arginine-rich (SR) proteins are involved in the pre-mRNA processing and releasing through nucleus into nucleoplasm. CLK1, which could phosphorylate the specific SR proteins (1) such as ASF/SF2, may directly regulate the activity and compartmentalization of SR splicing factors (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CLK1, 10ug (Human, recombinant; amino acids 129-end). MW: ~66kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. CLK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1195/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.