Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Wizard Plus SV Minipreps Start-Up Kit, 110V Electrical (750 preps, manifold and free vacuum pump), 1 each
- Size:1 each
- Storage Conditions:15°C to 30°C
- Description:Wizard Plus SV Minipreps Start-Up Kit
- Cat. No.:PAA6760
- Supplier no.:A6760
Specifications
About this item
Bundle of 750 minipreps, vacuum manifold and free vacuum pump. Wizard Plus SV Minipreps is silica membrane-based system for simple, rapid isolation of plasmid DNA from 1-10ml E. coli cultures in 45 minutes or less.
- Plasmid Miniprep System, Vacuum Manifold and Free Vacuum Pump
- 1-20microg high-quality plasmid DNA from 1-10ml cultures
- Process up to 20 minipreps in 45 minutes
- Choice of spin or vacuum purification formats
Bundle of 750 minipreps, vacuum manifold and free vacuum pump.The Wizard Plus SV Minipreps DNA Purification System, a silica membrane-based system, provides a simple and reliable method for rapid isolation of plasmid DNA. The entire miniprep procedure can be completed in 45 minutes or less, depending on the number of samples processed. Using the system, plasmid DNA can be purified from 1-10ml of overnight E. coli culture. The purified plasmid DNA can be used directly for automated fluorescent BigDye terminator DNA sequencing as well as for other standard molecular biology techniques without further manipulation. It also can be used for in vitro transcription reactions when supplemented with a ribonuclease inhibitor such as Recombinant RNasin Ribonuclease Inhibitor (Cat.# N2511).