Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Alkaline Protease

130 ml
Catalog # PAA1721
Supplier:   Promega Corporation
 :  A1721
Alkaline Protease
Catalog # PAA1721
CAS Number:  
Alkaline Protease
Alkaline Protease

130 ml

CAS Number:  
Price
$196.13
Log in for availability
Each (130ml)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Alkaline Protease (APA) is a component of the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751).

  • Reagent Used in DNA Isolation Protocols
  • Component of the ReliaPrep™ Large Volume HT gDNA Isolation System (Cat.# A2751) available as a standalone item

Alkaline Protease (APA) is a component of the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751).

Specifications

Specifications

  • Size:
    130 ml
  • Storage Conditions:
    15...30 °C
  • Enzyme Synonyms:
    APA
  • Cat. No.:
    PAA1721
  • Enzyme Name:
    Alkaline Protease

Specifications