Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard Genomic DNA Purification Kit, 100 isolations × 10ml, Promega®
Wizard Genomic DNA Purification Kit, 100 isolations × 10ml, Promega®
Catalog # PAA1620
CAS Number:  
undefined
Wizard Genomic DNA Purification Kit, 100 isolations × 10ml, Promega®
Catalog # PAA1620
Supplier Number:  A1620
CAS Number:  

Specifications

  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Biofluids (serum, plasma, saliva)
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Wizard Genomic DNA Purification Kit, 100 isolations x 10ml
  • Size:
    100 isolations x 10ml
  • Sample Size:
    Scalable
  • Description:
    Wizard Genomic DNA Purification Kit
  • Yield:
    Varies based on starting material.
  • Cat. No.:
    PAA1620
  • Supplier no.:
    A1620

Specifications

About this item

An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.

  • Manual, Solution-Based DNA Extraction Method
  • DNA from blood, animal and plant cells, bacteria or yeast
  • 60-minute, scalable protocol
  • Kit size based on number of isolations from whole blood, other sample types may vary

The Wizard Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridizations (e.g., Southern and dot/slot blots).