Order Entry
Puerto Rico
ContactUsLinkComponent
 
ADP-Glo Kinase Assay + BTK Kinase Enzyme System, 1 each, Promega
Catalog # PAV9071
CAS Number:  
undefined
ADP-Glo Kinase Assay + BTK Kinase Enzyme System, 1 each, Promega
Catalog # PAV9071
Supplier Number:  V9071
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + BTK Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9071
  • Supplier no.:
    V9071

Specifications

About this item

Recombinant full-length human BTK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. BTK (also known as Bruton tyrosine kinase) plays a crucial role in B-lymphocyte differentiation and activation.

  • Easily Screen and Profile BTK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human BTK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. BTK (also known as Bruton tyrosine kinase) plays a crucial role in B-lymphocyte differentiation and activation. BTK interacts with SRC homology 3 domains of FYN, LYN and HCK that are activated upon stimulation of B- and T-cell receptors. Defects in the BTK gene cause Agammaglobulinemia, an X-linked immunodeficiency characterized by failure to produce mature B lymphocyte cells and associated with a failure of Ig heavy chain rearrangement. The unique role of BTK makes it a desirable target for potential anti-cancer, anti-inflammatory and anti-viral agents as well as other treatments. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: BTK, 10ug (Human, recombinant full-length). MW: ~78kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. BTK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/695/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.