Order Entry
Puerto Rico
ContactUsLinkComponent
 
GoTaq Probe 2-Step RT-qPCR System, 2ml, Promega®
Catalog # PAA6110
CAS Number:  
undefined
GoTaq Probe 2-Step RT-qPCR System, 2ml, Promega®
Catalog # PAA6110
Supplier Number:  A6110
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    RT-qPCR probe
  • Description:
    GoTaq Probe 2-Step RT-qPCR System
  • Cat. No.:
    PAA6110
  • No. of reactions:
    20 µl× 50 RT and 50 µl× 80 qPCR at 50 µl, 20 µl× 50 RT and 20 µl× 200 qPCR at 20 µl, 20 µl× 50 RT and 10 µl× 400 qPCR at 10 µl
  • Volume:
    2 ml
  • Supplier no.:
    A6110

Specifications

About this item

GoTaq Probe RT-qPCR Systems enable detection and relative quantification of RNA expression levels.

  • Robust System to Simplify RNA Expression Analysis
  • Integrated components for reverse transcription and downstream qPCR, including RNasin(R) Ribonuclease Inhibitor
  • Compatible with both fast and standard cycling methods using probe-based assays
  • Sensitive detection for quantitation over a broad dynamic range

The GoTaq Probe 2-Step RT-qPCR System facilitates detection and relative quantification of RNA expression levels via a two-step RT-qPCR method using integrated components: GoScript Reverse Transcription System and GoTaq Probe qPCR Master Mix. The GoScript Reverse Transcription System includes an optimized reaction buffer and M-MLV reverse transcriptase designed to enable efficient synthesis of first-strand cDNA in preparation for PCR amplification. The cDNA product can be added directly to downstream qPCR amplification reactions. The GoTaq Probe 1-Step RT-qPCR System enables detection and relative quantification of RNA expression levels using a one-step RT-qPCR method, combining GoScript Reverse Transcriptase and GoTaq Probe qPCR Master Mix in a single-step real-time amplification reaction.

Caution: Store all components between –30°C and –10°C. Protect components from light at all times. For best results, mix thawed solutions gently to minimize aeration and foaming, and keep on ice. For short–term storage and frequent use, the GoTaq qPCR Master Mix, 2X, may be kept at 2 to 8°C for up to three months if protected from light.