To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:qPCR probe
- Description:GoTaq Probe qPCR Master Mix
- Environmentally Preferable:
- Cat. No.:PAA6101
- No. of reactions:80 at 50 µl, 200 at 20 µl, 400 at 10 µl
- Volume:2 ml
- Supplier no.:A6101
Specifications
About this item
GoTaq Probe qPCR Master Mix is a ready-to-use stabilized 2X formulation containing GoTaq Hot Start Polymerase, MgCl2, dNTPs and proprietary reaction buffer.
- Convenient qPCR Amplification Using Hydrolysis Detection Probes
- Ready-to-use mix of optimized components for robust amplification
- Compatible with both fast and standard cycling methods
- Sensitive detection for quantitation over a broad dynamic range
The GoTaq Probe qPCR Master Mix is optimized for fast and reproducible quantitative PCR assays in the hydrolysis probe detection format. The master mix is provided as a ready-to-use, stabilized 2X formulation and contains GoTaq Hot Start Polymerase, MgCl2, dNTPs and a proprietary reaction buffer. This master mix does not contain a reference dye; a separate tube of CXR reference dye is included.