Order Entry
Puerto Rico
ContactUsLinkComponent
ADP-Glo Kinase Assay + AMPK (A2/B1/G1) Kinase Enzyme System, 1 each, Promega
ADP-Glo Kinase Assay + AMPK (A2/B1/G1) Kinase Enzyme System, 1 each, Promega
Catalog # PAV4015
CAS Number:  
undefined
ADP-Glo Kinase Assay + AMPK (A2/B1/G1) Kinase Enzyme System, 1 each, Promega
Catalog # PAV4015
Supplier Number:  V4015
CAS Number:  

Specifications

  • Description:
    ADP-Glo Kinase Assay + AMPK (A2/B1/G1) Kinase Enzyme System, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV4015
  • Supplier no.:
    V4015

Specifications

About this item

Recombinant full-length human AMPK (A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for diabetes treatment.

  • Easily Screen and Profile AMPK (A2/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Recombinant full-length human AMPK (combination of A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for the treatment of diabetes (1). AMPK is viewed as a fuel sensor for glucose and lipid metabolism by modulating the activity of the autonomous nervous system in vivo. Short-term overexpression of a constitutively active form of AMPK in the liver leads to mild hypoglycemia and fatty liver due to increased fatty acid utilization (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AMPK (A2/B1/G1), 10ug (Human, recombinant full-length). MW: ~69kDa (A2); ~38kDa (B1); ~40kDa (G1). Substrate: SAMStide (HMRSAMSGLHLVKRR); derived from the mouse acetyl-Coenzyme A carboxylase alpha (amino acids 73-85). Other: Reaction Buffer, DTT, AMP Solution. AMPK (A2/B1/G1) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5563/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.