Order Entry
Puerto Rico
ContactUsLinkComponent
ReliaPrep Blood gDNA Miniprep System, Promega®
ReliaPrep Blood gDNA Miniprep System, Promega®
Certificates
undefined
ReliaPrep Blood gDNA Miniprep System, Promega®
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Storage Conditions:
    15°C to 30°C
  • Description:
    ReliaPrep Blood gDNA Miniprep System

Specifications

Related Information