Order Entry
Puerto Rico
ContactUsLinkComponent
ReliaPrep Large Volume HT gDNA Isolation System, 1 each, Promega
ReliaPrep Large Volume HT gDNA Isolation System, 1 each, Promega
Catalog # PAA2751
CAS Number:  
undefined
ReliaPrep Large Volume HT gDNA Isolation System, 1 each, Promega
Catalog # PAA2751
Supplier Number:  A2751
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    ReliaPrep Large Volume HT gDNA Isolation System, 1 each
  • Size:
    1 each
  • Storage Conditions:
    15°C to 30°C
  • Description:
    ReliaPrep Large Volume HT gDNA Isolation System
  • Cat. No.:
    PAA2751
  • Supplier no.:
    A2751

Specifications

About this item

The ReliaPrep Large Volume HT gDNA Isolation System isolates genomic DNA (gDNA) from 1-10ml of blood in a scalable format. The chemistry eliminates tedious centrifugation steps as well as the use of hazardous chemicals.

  • Automated DNA Isolation from 1-10ml Blood
  • Effective recovery from pristine, hemolysed or frozen samples
  • Purified DNA ready for use in downstream assays or archiving
  • Chemistry scaled for each sample, reducing waste

The ReliaPrep Large Volume HT gDNA Isolation System isolates genomic DNA (gDNA) from 1-10ml of blood in a scalable format. The chemistry eliminates tedious centrifugation steps as well as the use of hazardous chemicals, which are inherent in precipitation-based chemistries. Each reagent kit provides enough reagents to process up to 96 x 10ml whole blood samples. The system has been automated on robotic liquid-handling workstations, allowing walkaway purification of genomic DNA from 1-10ml of whole blood, regardless of sample storage or shipping conditions. For low-throughput isolation of gDNA from up to 32 samples at one time, the HSM 2.0 can be used in a manual mode, where the user performs the pipetting functions. The HSM has software that controls the instrument and directs the user through the purification protocol.