Order Entry
Puerto Rico
ContactUsLinkComponent
 
Bottle for 50% Ethanol, 1 each, Promega
  PAA2691
undefined
Bottle for 50% Ethanol, 1 each, Promega
  PAA2691
 :  A2691

 

  • Description:
    Bottle for 50% Ethanol, 1 each
  • Cat. No.:
    PAA2691
  • Supplier no.:
    A2691

 

 

Reagent bottle for ethanol solution used in automated purification of DNA from 1 to 10 ml blood on liquid-handling systems.

Accessory for use with the ReliaPrep™ large volume HT gDNA isolation system.