Order Entry
Puerto Rico
ContactUsLinkComponent
 
PureYield Plasmid Maxiprep System, Promega
 
undefined
PureYield Plasmid Maxiprep System, Promega
 

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Storage Conditions:
    15°C to 30°C
  • Description:
    PureYield Plasmid Maxiprep System