ReliaPrep FFPE gDNA Miniprep System, Promega
Supplier: Promega Corporation
Certificates
About this item
The ReliaPrep FFPE gDNA Miniprep System provides a complete, all-inclusive method for purifying quality genomic DNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.
- Intact, Amplifiable DNA from FFPE Tissue
- Minimal preparation time: simply add ethanol and go!
- Safely deparaffinize without using harsh organic solvents
- No overnight digestion required
The ReliaPrep FFPE gDNA Miniprep System provides a complete, all-inclusive method for purifying quality genomic DNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion. Genomic DNA can be isolated from FFPE tissue in approximately two and one-half hours with minimal hands-on time.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:FFPE tissues
- Target molecule:DNA
- Storage Conditions:15°C to 30°C
- Description:ReliaPrep FFPE gDNA Miniprep System