Order Entry
Puerto Rico
ContactUsLinkComponent
 
PureYield™ Plasmid Maxiprep Start-Up Kit, Promega®
  PAA6730
 :  
undefined
PureYield™ Plasmid Maxiprep Start-Up Kit, Promega®
  PAA6730
 :  A6730
 :  

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    PureYield Plasmid Maxiprep Start-Up Kit, 110V Electrical (50 preps, manifold, 4 elution devices and free vacuum pump)
  • Size:
    1 each
  • Storage Conditions:
    15 °C to 30 °C
  • Description:
    PureYield Plasmid Maxiprep Start-Up Kit
  • Cat. No.:
    PAA6730
  • Supplier no.:
    A6730

 

 

The PureYield™ Plasmid Maxiprep System provides a rapid method for purification of up to 1 mg of transfection-quality plasmid DNA from 250 ml bacterial cultures. The system is designed for use with a vacuum source and vacuum manifold, greatly reducing the time spent on purification compared to silica resin or other membrane-based methods.

  • Up to 1 mg of plasmid from 250 ml culture
  • Highly pure, concentrated plasmid DNA
  • No tedious, gravity-drip columns or post-elution alcohol precipitation

PureYield™ Plasmid Maxipreps include a unique Endotoxin Removal Wash designed to remove substantial amounts of protein, RNA and endotoxin contaminants from purified plasmid DNA. Removal of contaminants improves performance in sensitive applications such as eukaryotic transfection, in vitro transcription and transcription/translation applications. Purification is achieved without isopropanol precipitation of purified plasmid DNA.