Order Entry
United States
ContactUsLinkComponent
 
Wizard SV 9600 Plasmid DNA Purification System, Promega
Catalog # PAA2258
CAS Number:  
undefined
Wizard SV 9600 Plasmid DNA Purification System, Promega
Catalog # PAA2258
Supplier Number:  A2258
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Wizard SV 9600 Plasmid DNA Purification System, 100 x 96 preps
  • Size:
    100 x 96 preps
  • Environmentally Preferable:
  • Storage Conditions:
    15°C to 30°C
  • Description:
    Wizard SV 9600 Plasmid DNA Purification System
  • Cat. No.:
    PAA2258
  • Supplier no.:
    A2258

Specifications

About this item

The Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format.

  • Silica Membrane-Based, 96-Well, High-Throughput Plasmid Purification
  • 60-minute protocol
  • Manual and automated formats
  • Spin or vacuum-based methods

The Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format. A single plate can be processed in 60 minutes or less. The purified plasmid can be used directly for automated fluorescent DNA sequencing as well as for other standard molecular biology techniques, including restriction enzyme digestion. The Wizard SV 96 and SV 9600 Systems are designed for use either in a manual format or with Beckman Coulter or PerkinElmer automated instruments.