ContactUsLinkComponent
200 bp DNA Step Ladder, 100 µg, Promega
200 bp DNA Step Ladder, 100 µg, Promega
  PAG6961
 :  
undefined
200 bp DNA Step Ladder, 100 µg, Promega
  PAG6961
 :  G6961
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    200bp DNA Step Ladder, 100microg
  • Size:
    100 μg
  • Environmentally Preferable:
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAG6961
  • Supplier no.:
    G6961
Thirty-three blunt-ended DNA fragments ranging from 200bp to 6,600bp in 200bp increments. The 1,000bp band appears more intense than all other bands.
  • Thirty-Three Blunt-Ended DNA Fragments of 200-6,600bp in 200bp Increments
  • Load 1microl/lane
  • Supplied at a concentration of 1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 200bp DNA Step Ladder: Thirty-three blunt-ended DNA fragments ranging from 200bp to 6,600bp in 200bp increments. The 1,000bp band appears more intense than all other bands, which are of approximately equal intensity. Recommended loading: 1microl/lane.