About this item
The 10X Flexi Enzyme Blend (SgfI and PmeI) is required to screen Flexi Vector clones for the presence of the protein-coding region of interest. This enzyme blend can also be used to cleave PCR products prior to cloning into the Flexi Vectors or transfer p
- Reagent Used with Flexi(R) Cloning System
- Required to screen Flexi(R) Vector clones for presence of protein-coding region of interest
- Also can be used to cleave PCR products prior to cloning into Flexi(R) Vectors or transfer protein-coding regions between Flexi(R) Vectors
The 10X Flexi Enzyme Blend (SgfI and PmeI) is required to screen Flexi Vector clones for the presence of the protein-coding region of interest. This enzyme blend can also be used to cleave PCR products prior to cloning into the Flexi Vectors or transfer protein-coding regions between Flexi Vectors. For more information see the Flexi Vector Systems Technical Manual #TM254. Recognition Sites: SgfI recognition site: 5…GCGATtCGC…3 3…CGCsTAGCG…5. PmeI recognition site: 5…GTTTtAAAC…3 3…CAAAsTTTG…5. Storage Buffer: The 10X Flexi Enzyme Blend (SgfI and PmeI) is supplied in 10mM Tris-HCl (pH 7.4 at 25°C), 150mM KCl, 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.35mg/ml BSA, 0.05% Thesit and 50% glycerol. 5X Flexi Digest Buffer: The composition of 5X Flexi Digest Buffer is 50mM Tris-HCl (pH 7.9 at 37°C), 50mM MgCl2, 250mM NaCl, 5mM DTT, 0.5mg/ml acetylated BSA.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage Conditions:–30...–10 °C
- Enzyme Name:10X Flexi Enzyme Blend (SgfI & PmeI)