Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard MagneSil Sequencing Reaction Clean-Up System, Promega
Wizard MagneSil Sequencing Reaction Clean-Up System, Promega
Certificates
undefined
Wizard MagneSil Sequencing Reaction Clean-Up System, Promega

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Storage Conditions:
    15°C to 30°C
  • Description:
    Wizard MagneSil Sequencing Reaction Clean-Up System

Specifications