Order Entry
Puerto Rico
ContactUsLinkComponent
BCA Protein Assay kit (BCA-PAK), OZ Biosciences
BCA Protein Assay kit (BCA-PAK), OZ Biosciences
Catalog # 103549-592
Supplier:  OZ Biosciences
CAS Number:  
undefined
BCA Protein Assay kit (BCA-PAK), OZ Biosciences
Catalog # 103549-592
Supplier:  OZ Biosciences
Supplier Number:  BCA2500
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BCA Protein Assay Kit
  • Brand Name:
    OZBI
  • No. of transfections/assays:
    2500 assays
  • Cat. No.:
    103549-592
  • Volume:
    500 ml BCA reagent A + 15 ml copper solution B + × 1 ml - 1500 µg/ml BSA

Specifications

About this item

BCA Protein Assay kit (BCA-PAK) is useful for colorimetric detection and quantification of total protein content even in the presence of detergents.

This kit is based on the reduction of Cu2+ to Cu1+ by protein in alkaline solution; monovalent copper ions produced are detected in a concentration-dependent manner. Bicinchoninic acid (BCA) chelates with the reduced copper Cu1+ and forms a water-soluble purple reaction complex that exhibits a strong absorbance at 562 nm. Absorbance is linear over a wide range of protein concentrations between 25-2000 µg/mL. In general, protein concentrations are estimated with reference to a commonly used protein standard; the kit also includes Bovine Serum Albumin (BSA) at 1.5 mg/mL as a protein standard for a convenient preparation of protein concentration standard curves.      

Delivery: Shipped at room temperature.
Packaging: Store at 4°C