HYPE-293 Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
HYPE-293™ Transfection Kit is dedicated to achieve High Yield Protein Expression in HEK-293, 293-S, 293-F or any 293-related cells growing in suspension.
- Reach optimal Protein & Antibody production yield
- Suitable for all HEK293 cells growing in suspension
- Compatible with any synthetic or regular media used for protein production
- Ideal for bioreactor, spinner, or flasks
- Animal origin free
Scale-up to larger volumes for production of milligrams of protein per liter of cell culture is straightforward and easy with simple and cost efficient handling steps.
Delivery information: Shipped at room temperature.
Packaging: Store Hype-293 Reagent –20 °C - B293 reagent –20 °C.
Specifications
- Bioproduction:Yes
- Cell Specific:HEK293 suspension