ContactUsLinkComponent
MetriZyme™ Dual Enzymatic Instrument Detergent, Metrex
MetriZyme™ Dual Enzymatic Instrument Detergent, Metrex
Catalog # 89132-869
Supplier:  Metrex
MetriZyme™ Dual Enzymatic Instrument Detergent, Metrex
Catalog # 89132-869
Supplier:  Metrex
Supplier Number:  10-4000
Specifications
  • Description:
    Dual Enzymatic Instrument Detergent
  • Cat. No.:
    89132-869
  • Packaging:
    Jug
  • Volume:
    1 Gal.
  • Supplier no.:
    10-4000
Specifications
About this item
MetriZyme™ is designed for superior performance in the presence of high organic debris while offering the broadest cleaning action on a variety of protein soils
  • Powerful cleaning
  • Ready to use
  • Liquid concentrate
Dual enzymatic instrument detergent includes an enzyme stabilizer, a buffer to maintain pH, a preservative, an antifoaming agent and solution dye to differentiate it from water and other solutions.
Surfactants provide additional cleaning on carbohydrates, lipids and protein soils.