Order Entry
Puerto Rico
ContactUsLinkComponent
POLYVIEW® PLUS AP (anti-mouse) reagent, Enzo Life Sciences
POLYVIEW® PLUS AP (anti-mouse) reagent, Enzo Life Sciences
Catalog # 76201-850
Supplier:  Enzo Life Sciences
CAS Number:  
POLYVIEW® PLUS AP (anti-mouse) reagent, Enzo Life Sciences
Catalog # 76201-850
Supplier:  Enzo Life Sciences
Supplier Number:  ACC114-0150
CAS Number:  

Specifications

  • Conjugation:
    AP (Alkaline Phosphatase)
  • Description:
    AP (Anti-Mouse) Reagent POLYVIEW® Plus
  • Size:
    150 tests
  • ImmunoChemistry:
    Yes
  • Antibody Type:
    Secondary
  • Storage Temperature:
    +4°C
  • Shipping Temperature:
    Blue Ice Not Frozen
  • Cat. No.:
    76201-850

Specifications

About this item

Recommended Applications: IHC, ISH (in situ hybridization)

  • Ready-to-use reagent
  • Biotin-free nanopolymer detection circumvents endogenous biotin background
  • High intensity color development delivers sharp, crisp staining
  • Compatible with both automated and manual platforms
  • The Antibody Blocker/Diluent further reduces background (sold separately)
  • Customize with HIGHDEF® chromogens of your choice (sold separately)

The POLYVIEW® PLUS AP (anti-mouse) reagent is a ready-to-use, biotin-free, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW® PLUS AP (anti-mouse) reagent can be adapted for in situ hybridization detection.