Accessories for GloMax® Discover and Explorer Systems., Promega
Supplier: Promega Corporation
Certificates
About this item
These accessories are intended for use with the GloMax Discover and Explorer Systems.
- For use with GloMax(R) Discover and Explorer Systems
- Filter Paddles only available for GloMax(R) Discover System
- Absorbance Module only available for GloMax(R) Explorer System
These accessories are intended for use with the GloMax Discover and Explorer Systems.