Human Recombinant Pd-L1
76202-250
:
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Endotoxin Content:<0.1 EU/μG (LAL test, Cape Cod Associates)
- Protein/Peptide Name:Pd-L1
- Purity:>90% (SDS-PAGE)
- Molecular Weight:~27kDa (actual is a doublet at 45kDa and 50kDa)
- Concentration:0.5 mg/mL after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Cat. No.:76202-250
Produced in HEK cells. Human PD-L1 (aa 19-239) is fused at the N-terminus to a His-tag and FLAG®-tag.