ContactUsLinkComponent
MagnaBot II Magnetic Separation Device, Promega
MagnaBot II Magnetic Separation Device, Promega
  PAV8351
undefined
MagnaBot II Magnetic Separation Device, Promega
  PAV8351
 :  V8351
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    MagnaBot II Magnetic Separation Device, 1 each
  • Size:
    1 each
  • Storage Temperature:
    15 - 30 °C
  • Cat. No.:
    PAV8351
  • Supplier no.:
    V8351
The MagnaBot II Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles with a 96-well PCR plate.
  • Designed to work with a 96-well PCR plate
  • Allows the quantitative removal of liquids
The MagnaBot II Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles with a 96-well PCR plate. A 96-well PCR plate containing MagneSil Paramagnetic Particles is placed on the unit to draw the particles to the side and away from the bottom of each well. This allows the quantitative removal of liquids.