ContactUsLinkComponent
MagnaBot 96 Magnetic Separation Device, Promega
MagnaBot 96 Magnetic Separation Device, Promega
Catalog # PAV8151
undefined
MagnaBot 96 Magnetic Separation Device, Promega
Catalog # PAV8151
Supplier Number:  V8151
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MagnaBot 96 Magnetic Separation Device, 1 each
  • Size:
    1 each
  • Storage Temperature:
    15 - 30 °C
  • Cat. No.:
    PAV8151
  • Fits:
    Wizard® Magnetic DNA purification systems
  • Supplier no.:
    V8151
Specifications
About this item
The MagnaBot 96 Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles.
  • Designed for high-throughput bioseparation using magnetic particles
  • Compatible with many plates
The MagnaBot 96 Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles, which use the principle of magnetic separation as an alternative to vacuum filtration and centrifugation separation formats. This device is compatible with Collection Plates (Cat.# A9161), 1.2ml, Round-Bottom Deep Well Plates (Cat.# V6771) and Greiner 96-well P.S. V-bottom plates (Greiner Cat.# 651101). A generic 96-well plate may not be compatible.