Deep Well MagnaBot 96 Magnetic Separation Device, Promega
PAV3031
- Description:Deep Well MagnaBot 96 Magnetic Separation Device, 1 each
- Size:1 each
- Storage Temperature:15 - 30 °C
- Cat. No.:PAV3031
- Supplier no.:V3031
The Deep Well MagnaBot(R) 96 Magnetic Separation Device is designed for high-throughput bioseparation in deep-well plates using magnetic particles such as MagneSil(R) Paramagnetic Particles.
- Designed for high-throughput bioseparation in deep-well plates using magnetic particles
- An alternative to vacuum filtration and centrifugation separation formats
- Use for automated nucleic acid purification applications
The Deep Well MagnaBot(R) 96 Magnetic Separation Device is designed for high-throughput bioseparation in deep-well plates using magnetic particles such as MagneSil(R) Paramagnetic Particles. Use as an alternative to vacuum filtration and centrifugation separation formats. Use for automated nucleic acid purification applications such as plasmid purification, PCR clean-up and dye terminator clean-up prior to automated sequencing.