Order Entry
Puerto Rico
ContactUsLinkComponent
CheckMate Negative Control Vectors, 1 set, Promega
CheckMate Negative Control Vectors, 1 set, Promega
Catalog # PAC9380
CAS Number:  
undefined
CheckMate Negative Control Vectors, 1 set, Promega
Catalog # PAC9380
Supplier Number:  C9380
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    CheckMate™ Negative Control Vectors
  • Size:
    1 set
  • Cat. No.:
    PAC9380
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    C9380

Specifications

About this item

Powerful system for detecting protein:protein interactions in vivo.

  • Powerful System for Detecting Protein:Protein Interactions In Vivo
  • Mammalian system for studying interactions in cell line of choice
  • Study post-translational modifications including glycosylation, phosphorylation and acylation

Two-hybrid systems are extremely powerful methods for detecting protein:protein interactions in vivo. The basis of two-hybrid systems is the modular domains found in some transcription factors: a DNA-binding domain, which binds to a specific DNA sequence, and a transcriptional activation domain, which interacts with the basal transcriptional machinery. A transcriptional activation domain in association with a DNA-binding domain will promote the assembly of RNA polymerase II complexes at the TATA box and increase transcription. In the CheckMate Mammalian Two-Hybrid System the DNA-binding domain and the transcriptional activation domain, produced by separate plasmids, are closely associated when one protein (X) fused to a DNA-binding domain interacts with a second protein (Y) fused to a transcriptional activation domain. In this system, interaction between proteins X and Y results in transcription of a reporter gene.