Order Entry
Puerto Rico
ContactUsLinkComponent
pF5K CMV-neo Flexi Vector, 20 µg, Promega
pF5K CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAC9411
CAS Number:  
undefined
pF5K CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAC9411
Supplier Number:  C9411
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    pF5K CMV-neo Flexi® Vector
  • Size:
    20 μg
  • Cat. No.:
    PAC9411
  • Storage temperature:
    –30...–10 °C
  • Embl accession no.:
    DQ487156
  • Supplier no.:
    C9411

Specifications

About this item

The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences.

  • Untagged Proteins Expressed in Mammalian Cells with a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity

The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. The pF5A and pF5K CMV-neo Flexi Vectors below are designed specifically for high-level expression of proteins in mammalian cells from the CMV promoter with a selectable marker. All inserts may be confirmed by cell-free expression with the TNT T7 Quick System (Cat.# L1170). Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.