Order Entry
Puerto Rico
ContactUsLinkComponent
pF4K CMV Flexi Vector, 20 µg, Promega
pF4K CMV Flexi Vector, 20 µg, Promega
Catalog # PAC8491
CAS Number:  
undefined
pF4K CMV Flexi Vector, 20 µg, Promega
Catalog # PAC8491
Supplier Number:  C8491
CAS Number:  

Specifications

  • Description:
    pF4K CMV Flexi® Vector
  • Size:
    20 μg
  • Cat. No.:
    PAC8491
  • Storage temperature:
    –30...–10 °C
  • Embl accession no.:
    AY753781
  • Supplier no.:
    C8491

Specifications

About this item

Designed for high-level expression of proteins in mammalian cells from the CMV promoter without a selectable marker

  • Untagged Proteins Expressed in Mammalian Cells Without a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity

The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. The pF4A and pF4K CMV Flexi Vectors below are designed specifically for high-level expression of proteins in mammalian cells from the CMV promoter without a selectable marker. All inserts may be confirmed by cell-free expression with the TNT T7 Quick System (Cat.# L1170). Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.