Order Entry
Puerto Rico
ContactUsLinkComponent
 
pGEM-7Zf(+) Vector, 20 µg, Promega
Catalog # PAP2251
CAS Number:  
undefined
pGEM-7Zf(+) Vector, 20 µg, Promega
Catalog # PAP2251
Supplier Number:  P2251
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAP2251
  • Storage temperature:
    Less than –65 °C
  • Supplier no.:
    P2251

Specifications

About this item

Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.

  • Standard Cloning Vectors and Templates for In vitro Transcription and Circular ssDNA Production
  • Blue/white screening for easily identifying recombinant clones
  • In vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Orientation of the f1 origin differentiates vectors (+ and -)

The pGEM-7Zf(+) and pGEM-7Zf(-) Vectors are derivatives of the pGEM-3Zf(+) Vector and contain the origin of replication of the filamentous phage f1. These plasmids serve as standard cloning vectors, as templates for in vitro transcription and can be used for the production of circular ssDNA. These plasmids contain SP6 and T7 RNA polymerase promoters flanking a region of multiple cloning sites within the alpha-peptide coding region of beta-galactosidase. Insertional inactivation of the alpha-peptide allows recombinant clones to be identified directly by color screening on indicator plates when using appropriate E. coli strains (e.g., JM109). The multiple cloning region is unique and includes restriction sites for ApaI, AatII, SphI, XbaI, XhoI, EcoRI, KpnI, SmaI, ClaI, HindIII, BamHI, SacI, BstXI and NsiI. This arrangement is designed specifically for generating unidirectional deletions with the Erase-a-Base System. pGEM-7Zf(+) and pGEM-7Zf(-) Vectors are identical except for the orientation of the f1 origin.