Adenylate Cyclase Toxin Increases Intracellular Camp Levels
76202-260
:
- Size:50 µg
- Endotoxin Content:<130 EU/mg (LAL method)
- Protein/Peptide Name:Adenylate Cyclase Toxin Increases Intracellular Camp Levels
- Purity:>80% (SDS-PAGE)
- Molecular Weight:~200 kDa
- Formulation:Liquid. In 0.05 M Tris, 8 M Urea, 0.002 M EDTA, pH 8.0.
- Cat. No.:76202-260
Increases intracellular cAMP levels
Produced in E. coli.