ContactUsLinkComponent
pSP73 Vector, 20 µg, Promega
pSP73 Vector, 20 µg, Promega
  PAP2221
 :  
undefined
pSP73 Vector, 20 µg, Promega
  PAP2221
 :  P2221
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    20 μg
  • Cat. No.:
    PAP2221
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    P2221
Used as standard cloning vector or for transcription of RNA in vitro.
  • Standard Cloning Vector That Also Transcribes RNA In Vitro
  • Versatile vector that can be used for standard cloning and in vitro transcription
  • SP6 and T7 RNA polymerase promoters flank the multiple cloning region
  • Multiple cloning site provides a convenient selection of restriction sites for cloning
The pSP73 Vector can be used as a standard cloning vector and also can be used for transcription of RNA in vitro The pSP73 Vector contains the SP6 and T7 RNA polymerase promoters and a unique multiple cloning region, which includes restriction sites for BglII, EcoRV, ClaI, EcoRI, SacI, KpnI, SmaI, BamHI, XbaI, SalI, AccI, PstI, SphI, HindIII, PvuII and XhoI. The pSP72 and pSP73 Vectors are essentially identical except for the orientation of the multiple cloning site region.