- Size:20 μg
- Cat. No.:PAP2191
- Storage temperature:–30...–10 °C
- Supplier no.:P2191
Used as standard cloning vector or for transcription of RNA in vitro.
- Standard Cloning Vector Used to Transcribe RNA In Vitro
- Versatile vector that can be used for standard cloning and in vitro transcription
- SP6 and T7 RNA polymerase promoters flank the multiple cloning region
- Multiple cloning site provides a convenient selection of restriction sites for cloning
The pSP72 Vector can be used as a standard cloning vector and also can be used for transcription of RNA in vitro. The pSP72 Vector contains the SP6 and T7 RNA polymerase promoters flanking a unique multiple cloning region, which includes restriction sites for XhoI, PvuII, HindIII, SphI, PstI, SalI, AccI, XbaI, BamHI, SmaI, KpnI, SacI, EcoRI, ClaI, EcoRV and BglII. The pSP72 and pSP73 Vectors are essentially identical except for the orientation of the multiple cloning site region.