Order Entry
United States
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (human female genomic)
DNA (Deoxyribonucleic acid) (human female genomic)
Catalog # 76202-256
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
DNA (Deoxyribonucleic acid) (human female genomic)
Catalog # 76202-256
Supplier:  Enzo Life Sciences
Supplier Number:  GEN118-0100
CAS Number:  

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76202-256

Specifications

About this item

 :
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922