DNA (Deoxyribonucleic acid) (human male genomic)
Catalog # 76202-254
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:100 µg
- Cat. No.:76202-254
Specifications
About this item
:
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922