IgA (Canine) ELISA, Eagle Biosciences
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Canine IgA ELISA is designed to detect canine IgA in complex samples.
- Dynamic Range: 3.125 - 200 ng/ml
- Sensitivity: 156 ng/ml (sample dilution 1:50)
- Incubation: 2.5 hours
The Eagle Biosciences Canine (Dog) IgA ELISA Assay Kit is designed for the quantitative determination of canine (dog) IgA in complex samples (serum or other biological samples). The Dog IgA ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.
Specifications
Specifications
- Description:IgA (Canine) ELISA
- Size:1 kit
- Cat. No.:102993-438
- Primary antibody reactivity:Dog
- Target protein:IgA
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



