Order Entry
Puerto Rico
ContactUsLinkComponent
Serotonin ELISA, Eagle Biosciences, Inc.
Serotonin ELISA, Eagle Biosciences, Inc.
Catalog # 102994-986
Supplier:  Eagle Biosciences
CAS Number:  
Serotonin ELISA, Eagle Biosciences, Inc.
Catalog # 102994-986
Supplier:  Eagle Biosciences
Supplier Number:  EA602/96
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    Serotonin
  • Description:
    Serotonin ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-986
  • Supplier no.:
    SER39-K01

Specifications

About this item

Serotonin ELISA detects the concentration of serotonin in Serum, Plasma, and Urine.

  • Dynamic Range: 15 - 2500 ng/mL
  • Sensitivity: 5 ng/mL
  • Incubation: 2.5 hours

Serotonin (5-Hydroxytryptamine), a biogenic amine, is a product of the tryptophan metabolism. It is a well evaluated neurotransmitter of the central nervous system and can be found in high concentrations in the chromaffine cells of the intestinal mucosa, in the platelets and the serotonergic neurones of the brain. Central-serotonergic neurons influence physiological functions such as sleep and the hormonal and cardio-vascular regulation. Increased serum levels can be found with malignant carcinoid, endogenous depression and schizophrenia.