About this item
Rat Albumin ELISA is to detect albumin in rat urine samples.
- Dynamic Range: 10 to 10240 ng/ml
- Sensitivity: 0.4 µg/ml
- Incubation: 2 hours
This kit is designed for the quantitative determination of albumin in rat urine samples.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Primary antibody reactivity:Rat
- Target protein:Albumin