Order Entry
Puerto Rico
ContactUsLinkComponent
Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences, Inc.
Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences, Inc.
Catalog # 102993-888
Supplier:  Eagle Biosciences
CAS Number:  
Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences, Inc.
Catalog # 102993-888
Supplier:  Eagle Biosciences
Supplier Number:  EA632/96
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    Adrenaline (Epinephrine)
  • Description:
    Adrenaline (Epinephrine) High Sensitive ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-888
  • Supplier no.:
    ADU39-K01

Specifications

About this item

Adrenaline Sensitive ELISA is intended to detect Adrenaline in biological fluids by a microplate .

  • Dynamic Range: 0.15 - 25 ng/ml
  • Sensitivity: 1.6 pg/mL
  • Incubation: overnight

Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.