Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Primary antibody reactivity:Pig
- Target protein:IFN gamma
- Description:Swine IFN-γ ELISA KIT
- Size:1 kit
- Cat. No.:102993-430
- Supplier no.:IFG51-K01
Specifications
About this item
Swine IFN Gamma EIA detect Swine Interferon Gamma concentrations in cell culture, serum, and plasma.
- Dynamic Range: 15.625 - 1000 pg/ml
- Sensitivity: 7 pg/ml
- Incubation: 3.0 hours
Interferon-gamma (IFN-g, also known as type II interferon) is an important immunoregulatory cytokine that was originally identified because of its anti-viral activity (1). It plays key roles in host defense by exerting antiviral, antiproliferative and immunoregulatory activities (2 - 5). On many cell types, IFN-g induces the production of cytokines and upregulates the expression of various membrane proteins including class I and II MHC Antigenss, Fc receptors, leukocyte adhesion molecules and B7 family Antigenss. IFN-g is a potent activator of macrophage effector functions. It potentiates the secretion of immunoglobulins by B cells, and directs the synthesis of IgG. IFN-g also influences T-helper cell phenotype development by inhibiting Th2 differentiation and stimulating Th1 development (2 - 5). Finally, IFN- promotes mononuclear cell chemotaxis by inducing the synthesis of CXCL9, CXCL10, CCL2, CCL3, CCL4, and CCL5(3). IFN-g is produced by a number of cell types, including dendritic epidermal/ T cells (6), keratinocytes (7), peripheral blood T cells (8), mast cells (9), neurons (10), CD8 T cells (11), macrophages (12), B cells (13), and neutrophils (14), NK cells (15), CD4 T cells (16) and testicular spermatids (17). The production of IFN-g is upregulated synergistically by IL-12, IL-18, IL-23 and IL-27 (18 - 21). Porcine IFN-g cDNA encodes a 166 amino acid (aa) residue precursor protein with a 20 aa signal sequence that is cleaved to generate a 146 aa residue mature IFN-g (22, 23). Porcine IFN-g is presumably a noncovalently linked homodimer (3). In the mature segment, porcine IFN-g shares 60%, 55%, 41%, 42%, 72%, and 72% aa sequence identity with human (24), guinea pig (25), mouse (26), rat (27), feline (28), and canine (29) IFN-g, respectively. The functional IFN-g receptor complex consists of two distinct subunits (30). The alpha subunit (IFN-gR1) binds IFN-g with high affinity and species specificity. The beta-subunit [IFN-gR2, also known as accessory factor-1 (AF-1)] interacts with the IFN- occupied-subunit in a species-specific manner and participates in JAK-STAT mediated signal transduction. Although the functional receptor is suggested to consist of homodimeric IFN-g in combination with two-chains, and two-chains (30, 31), it has been suggested that additional subunits may be involved (32, 33). Whereas the α-chain is expressed constitutively on many cell types, the cellular regulation of the β-chain correlates with an IFN-g responsive state and is tightly regulated (30).