Order Entry
Puerto Rico
ContactUsLinkComponent
S-Adenosyl-L-homocysteine (SAH) ELISA Assay Kit , Eagle Biosciences, Inc.
S-Adenosyl-L-homocysteine (SAH) ELISA Assay Kit , Eagle Biosciences, Inc.
Catalog # 102994-926
Supplier:  Eagle Biosciences
CAS Number:  
S-Adenosyl-L-homocysteine (SAH) ELISA Assay Kit , Eagle Biosciences, Inc.
Catalog # 102994-926
Supplier:  Eagle Biosciences
Supplier Number:  SAH31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    S-Adenosyl-L-homocysteine (SAH) ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-926
  • Supplier no.:
    SAH31-K01

Specifications

About this item

S-Adenosyl-L-homocysteine (SAH) ELISA is intended to detect SAH in serum or plasma.

  • Dynamic Range: 31.25 - 1000 nM
  • Sensitivity: 31.25 nM
  • Incubation: 1.5 hours

Methionine is an amino acid with sulfur methyl group, which is converted into S-adenosylmethionine (SAM) with ATP by methionine adenosyltransferase (EC 2.5.1.6). SAM is the sole methyl group donor for over 50 kinds of biologically active substances such as DNA, RNA, protein, phospholipids, hormones and neuro-transmitters, etc. After methyl group is transferred away from SAM under a mehtyltransferase, S-adenosylhomocysteine is formed, which is further metabolized into homocysteine (Hcy) after adenosine is removed. The homocysteine ​​is metabolized to methionine through N5-methyl tetrahydrofolate methyltransferase (EC1.1.1.68) and coenzyme vitamin B12 catalytic pathway accepting methyl from N5-methyltetrahydrofolate. This is methionine cycle.