Order Entry
Puerto Rico
ContactUsLinkComponent
Swine IL-10 ELISA KIT, Eagle Biosciences
Swine IL-10 ELISA KIT, Eagle Biosciences
Catalog # 102994-708
Supplier:  Eagle Biosciences
CAS Number:  
Swine IL-10 ELISA KIT, Eagle Biosciences
Catalog # 102994-708
Supplier:  Eagle Biosciences
Supplier Number:  L1051-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    Interleukin 10 (IL-10)
  • Description:
    Swine IL-10 ELISA KIT
  • Size:
    1 kit
  • Cat. No.:
    102994-708
  • Supplier no.:
    L1051-K01

Specifications

About this item

Swine IL-10 ELISA detects Swine Interleukin 10 concentrations in various samples.

  • Dynamic Range: 15.625 - 1000 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 3.0 hours

Interleukin 10, also known as cytokine synthesis inhibitory factor (CSIF), is the charter member of the IL-10 cytokine family. This family currently comprises IL-10, IL-19, IL-20, IL-22, IL-24, and IL-26/AK155 (1-4). All IL-10 family members are secreted α-helical proteins. Porcine IL-10 is a secreted, possibly glycosylated, polypeptide with an 18 kDa molecular weight (5). Based on human studies, porcine IL-10 is likely to circulate as a nondisulfide-linked homodimer (3). Porcine IL-10 is synthesized as a 175 amino acid (aa) precursor with an 18 aa signal sequence and a 157 aa mature form. The mature segment has one potential N-linked glycosylation site plus four cysteines which form two intrachain disulfide bridges (6). Mature porcine IL-10 shows 71%, 70%, 76%, 75%, 77%, and 71% aa sequence identity to rat (7), mouse (8), human (9), guinea pig (10), canine (11), and cotton rat (12) IL-10, respectively. Upon activation, mammalian cells known to secrete IL-10 include NK cells (13), cytotoxic CD8+ T cells secreting Th2-like cytokines (14), CD4+CD45RA- (memory) Th1 and Th2 cells (15), macrophages (16), monocytes (17), CD5+ and CD5- B cells (18, 19), dendritic cells (20, 21), hepatic stellate (Ito cells) (22), keratinocytes (23), melanoma cells (24), mast cells (25), placental cytotrophoblasts (26), and fetal erythroblasts (27).