To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:Mouse
- Description:Mouse Tissue-type Plasminogen Activator (tPA) Total Antigens ELISA
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:102995-046
- Supplier no.:TPN19-K01
Specifications
About this item
Mouse Tissue-type Plasminogen Activator (tPA) Total Antigen ELISA detects total tPA in mouse sample.
- Dynamic Range: 0.1 - 50 ng/ml
- Sensitivity: 0.1 ng/ml
- Incubation: 2 hours
The Mouse Tissue-type Plasminogen Activator (tPA) Total Antigens ELISA is intended for the quantitative determination of total plasminogen activator Antigens in biological fluids. Tissue plasminogen activator (tPA) is a serine protease that converts plasminogen to the active serine protease plasmin in the blood fibrinolytic system1,2. It also plays an important role in the removal of incipient thrombi3. tPA is widely used for the thrombolytic treatment of acute myocardial infarction3.