Order Entry
Puerto Rico
ContactUsLinkComponent
Anti tTg IgG (Tissue Transglutaminase) ELISA, Eagle Biosciences
Anti tTg IgG (Tissue Transglutaminase) ELISA, Eagle Biosciences
Catalog # 102995-022
Supplier:  Eagle Biosciences
CAS Number:  
Anti tTg IgG (Tissue Transglutaminase) ELISA, Eagle Biosciences
Catalog # 102995-022
Supplier:  Eagle Biosciences
Supplier Number:  4044
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Anti tTg IgG (Tissue Transglutaminase) ELISA
  • Size:
    1 kit
  • Cat. No.:
    102995-022
  • Supplier no.:
    TGG31-K01

Specifications

About this item

Anti-hu tTG IgG ELISA determines IgG autoantibodies to tissue transglutaminase in serum or plasma.

  • Dynamic Range: 1-300 U/mL
  • Sensitivity: 1U/mL
  • Incubation: 2.5 hours

Celiac disease, or gluten-sensitivity, is found already in neonates and is characterized by small intestinal damages leading to a so-called “flat” mucosa. Due to this extensive lesions mal-absorption occurs frequently accompanied with a depletion of key nutrients. Gliadin the alcohol soluble fraction of gluten represents the causative agent of celiac disease that provokes an inflammatory process in the small intestine. Gliadin is a substrate of tTG and cross-linked into high molecular complexes triggering probably both cellular and humoral immune responses. Incidence rates for celiac disease range from 1 in 300 (Western Ireland) to 1 in 4700 in European countries. However, a high number of subclinical cases of celiac disease have been detected by in-vitro tests revealing a prevalence of 4 in 1000. Individuals suffering from prolonged celiac disease additionally face an elevated risk of developing T cell lymphoma.Diagnosis of celiac disease comprises small intestine biopsy demonstrating a “flat” mucosa prior to a gluten-free diet and the following reconstitution of the mucosa after onset of the diet. Determination of anti-gliadin IgG and IgA by ELISA as well as the detection of anti-endomysium IgA by immunofluorescence has been considered as the main serological parameters for celiac disease so far.The identification of tissue transglutaminase as one of the main endomysial autoAntigenss and the availability of an easy to use and reliable ELISA kit employing recombinant human tissue transglutaminase pro¬mises the extension of diagnostic opportunities for celiac disease in future.IgA deficient celiac patients may demonstrate IgG autoantibodies to tTG only. Therefore, tTG IgG autoantibodies are the only serological parameter for celiac disease detectable in such patients apart from Gliadin IgG antibodies.