Order Entry
United States
ContactUsLinkComponent
Urinary Isoprostane ELISA Kit, Eagle Biosciences
Urinary Isoprostane ELISA Kit, Eagle Biosciences
Catalog # 102994-570
Supplier:  Eagle Biosciences
CAS Number:  
Urinary Isoprostane ELISA Kit, Eagle Biosciences
Catalog # 102994-570
Supplier:  Eagle Biosciences
Supplier Number:  ISO34-K01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugate ELISA:
    HRP
  • Host:
  • Primary antibody reactivity:
  • Description:
    Urinary Isoprostane EIA Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    102994-570
  • Supplier no.:
    ISO34-K01

Specifications

About this item

Urinary Isoprostane ELISA is quantitative determination of Isoprostane in human urine by microplate.

  • Dynamic Range: 0.05 - 100 ng/mL
  • Sensitivity: 0.05 ng/mL
  • Incubation: 2.5 hours

Isoprostanes are prostaglandin-like compounds that are produced by free radical mediated peroxidation of lipoproteins. This kit is for the quantification of 15-isoprostane F2t (also known as 8-epi-PGF2a or 8-iso-PGF2a) in urine samples. Levels of 15-isoprostane F2t in urine are useful for the non-invasive assessment of oxidant stress in vivo. 15-isoprostane F2t has also been shown to be a potent vasoconstrictor in rat kidneys and rabbit lungs, and plays a causative role in atherogenesis. Elevated isoprostane levels are associated with hepatorenal syndrome, rheumatoid arthritis, atherosclerosis, and carcinogenesis.