Order Entry
United States
ContactUsLinkComponent
Antibody to Cetuximab ELISA Assay Kit, Eagle Biosciences
Antibody to Cetuximab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-258
Supplier:  Eagle Biosciences
CAS Number:  
Antibody to Cetuximab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-258
Supplier:  Eagle Biosciences
Supplier Number:  IG-BB112
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    Antibody to Cetuximab ELISA Assay Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    103389-258

Specifications

About this item

Antibody to Cetuximab ELISA determines free antibodies to Cetuximab in serum and plasma.

  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours

The Eagle Biosciences Antibody to Cetuximab is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Cetuximab in serum and plasma samples. The Antibody to Cetuximab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.