Order Entry
Puerto Rico
ContactUsLinkComponent
RESISTIN ELISA, Eagle Biosciences, Inc.
RESISTIN ELISA, Eagle Biosciences, Inc.
Catalog # 102994-314
Supplier:  Eagle Biosciences
CAS Number:  
RESISTIN ELISA, Eagle Biosciences, Inc.
Catalog # 102994-314
Supplier:  Eagle Biosciences
Supplier Number:  E50
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    Resistin
  • Description:
    RESISTIN ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-314
  • Supplier no.:
    E50

Specifications

About this item

Resistin ELISA measures the amount of human resistin in serum and plasma

  • Dynamic Range: 20 - 1000 pg/mL
  • Sensitivity: 12 pg/ml
  • Incubation: 4 hours

Resistin, a cysteine-rich protein of 11.3 kDa (1), was firstly found in mice (2) and constitutes together with RELMα, RELMβ and RELMγ the protein family of resistin-like molecules (RELM).  In humans, resistin and RELMβ (1) but no other proteins of the RELM family were found. The human form of resistin shows a homology of 53% to the murine protein (4). It has 11 cysteine-residues, is synthesized as a propetide of 108 amino acids and secreted as a dimer, build by a disulfide bridge of cysteine residues (22). Beside this intermolecular disulfide bridge, 5 additional intramolecular ones exist (5,6).

For research use only.