Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugate ELISA:HRP
- Description:PROGRANULIN ELISA
- Size:1 kit
- Cat. No.:102994-296
- Supplier no.:E103
Specifications
About this item
Progranulin ELISA is to be used for the determination of Progranulin in serum or plasma samples.
- Dynamic Range: 75-2500 pg/mL
- Sensitivity: 18 pg/mL
- Incubation: 2 hours
Progranulin is also known as Granulin Epithelin Precursor, Proepithelin or Acrogranin. It is a 68.5 kDa protein, consisting of 593 amino acids (inclusive Signalpeptid), which appears in vivo in strongly glycosylated form and therefore has a size of approximately 90 kDa (1).